Läs mer
Benzarone, TRC
Produktkod. 30342307
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30342307 100 mg
1 options
Produktkod. 30342307 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB185150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzarone
CAS 1477-19-6
Molekylformel C17 H14 O3
Kvantitet 100 mg
Synonym (2-Ethyl-3-benzofuranyl)(4-hydroxyphenyl)methanone, (2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, 2-Ethyl-3-(4-hydroxybenzoyl)benzofuran, 2-Ethyl-3-(p-hydroxybenzoyl)benzofuran, 2-Ethyl-3-benzofuranyl p-hydroxyphenyl ketone, 2-Ethyl-4'-hydroxy-3-benzoylbenzofuran, Benzaron, Benzarone, Fagivil, Fragivil, Fragivix, L 2179-Labaz, L 2197, NSC 82134
InChI formel InChI=1S/C17H14O3/c1-2-14-16(13-5-3-4-6-15(13)20-14)17(19)11-7-9-12(18)10-8-11/h3-10,18H,2H2,1H3
LEDER CCc1oc2ccccc2c1C(=O)c3ccc(O)cc3
IUPAC-namn (2-ethyl-1-benzofuran-3-yl)-(4-hydroxyphenyl)methanone
Molekylvikt (g/mol) 266.29
Formel vikt 266.0943