Läs mer
Benzathine Ampicillin, TRC
Produktkod. 30343055
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30343055 50 mg
1 options
Produktkod. 30343055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB35662050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzathine Ampicillin
CAS 33276-75-4
Molekylformel C16H20N2 . 2(C16H19N3O4S)
Kvantitet 50 mg
Synonym N,N'-Dibenzylethylenediamine Ampicillin Disalt, Ampicillin Benzathine
InChI formel InChI=1S/2C16H19N3O4S.C16H20N2/c2*1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h2*3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);1-10,17-18H,11-14H2/t2*9-,10-,11+,14-;/m11./s1
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)O.CC4(C)S[C@@H]5[C@H](NC(=O)[C@H](N)c6ccccc6)C(=O)N5[C@H]4C(=O)O.C(CNCc7ccccc7)NCc8ccccc8
IUPAC-namn (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;N,N'-dibenzylethane-1,2-diamine
Molekylvikt (g/mol) 939.2
Formel vikt 938.3819