Läs mer
Benzene,1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)-] (>85%), TRC
Produktkod. 30329461
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30329461 100 mg
1 options
Produktkod. 30329461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB290020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzene,1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)-]
CAS 42757-55-1
Molekylformel C18H14Br8O4S
Kvantitet 100 mg
Synonym Benzene, 1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)- (9CI, ACI), 1,1'-Sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)benzene] (ACI), 1,3-Dibromo-5-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]sulfonyl-2-(2,3-dibromopropoxy)benzene, 4,4'-Bis(2,3-dibromopropoxy)-3,3',5,5'-tetrabromodiphenyl sulfone, Bis [3,5-dibromo-4-(2,3-dibromopropoxy) phenyl] sulfone, Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl] sulfone, DXFR 920, FCP 65CN, Fire Cut P 65CN, Flame Cut 161R, Nonnen 52, Nonnen PR 2, Nonnen PR 2H, PR 2, Tetrabromobisphenol S bis(2,3-dibromopropyl ether), XZ 6600
InChI formel InChI=1S/C18H14Br8O4S/c19-5-9(21)7-29-17-13(23)1-11(2-14(17)24)31(27,28)12-3-15(25)18(16(26)4-12)30-8-10(22)6-20/h1-4,9-10H,5-8H2
LEDER BrCC(Br)COc1c(Br)cc(cc1Br)S(=O)(=O)c2cc(Br)c(OCC(Br)CBr)c(Br)c2
IUPAC-namn 1,3-dibromo-5-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]sulfonyl-2-(2,3-dibromopropoxy)benzene
Molekylvikt (g/mol) 965.6
Formel vikt 957.408