missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Benzidine D8 500 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Kemiskt namn eller material | Benzidine ring-d8 |
| CAS | 92890-63-6 |
| Lösningsmedel eller matris | Acetonitrile/Methanol |
| Molekylformel | C12 2H8 H4 N2 |
| Kvantitet | 1 mL |
| Synonym | 4,4'-Diamino(biphenyl-d8) |
| InChI formel | InChI=1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2/i1D,2D,3D,4D,5D,6D,7D,8D |
| LEDER | [2H]c1c([2H])c(c([2H])c([2H])c1N)c2c([2H])c([2H])c(N)c([2H])c2[2H] |
| IUPAC-namn | 4-(4-amino-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuterioaniline |
| Molekylvikt (g/mol) | 192.29 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?