Läs mer
Benziodarone, TRC
Produktkod. 30329059
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30329059 10 mg
1 options
Produktkod. 30329059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB19786010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benziodarone
CAS 68-90-6
Molekylformel C17 H12 I2 O3
Kvantitet 10 mg
Synonym Methanone, (2-ethyl-3-benzofuranyl)(4-hydroxy-3,5-diiodophenyl)-, Ketone, 2-ethyl-3-benzofuranyl 4-hydroxy-3,5-diiodophenyl (6CI,7CI,8CI), 2-Ethyl-3-(3,5-diiodo-4-hydroxybenzoyl)-3-coumarone, 2-Ethyl-3-(3,5-diiodo-4-hydroxybenzoyl)benzofuran, 2-Ethyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran, 2-Ethyl-3-(4-hydroxy-3,5-diiodobenzoyl)coumarone, 2329 Labaz, Algocor, Amplivix, Benziodarone, Cardivix, Corofam, Coronal-Crinos, Dila-Vasal, Dilafurane, L 2329, NSC 82133, Retrangor
InChI formel InChI=1S/C17H12I2O3/c1-2-13-15(10-5-3-4-6-14(10)22-13)16(20)9-7-11(18)17(21)12(19)8-9/h3-8,21H,2H2,1H3
LEDER CCc1oc2ccccc2c1C(=O)c3cc(I)c(O)c(I)c3
IUPAC-namn (2-ethyl-1-benzofuran-3-yl)-(4-hydroxy-3,5-diiodophenyl)methanone
Molekylvikt (g/mol) 518.08
Formel vikt 517.8876