Läs mer
Benzo(b)chrysene, TRC
Produktkod. 30330411
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30330411 5 mg
1 options
Produktkod. 30330411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2170005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzo(b)chrysene
CAS 214-17-5
Molekylformel C22 H14
Kvantitet 5 mg
Synonym Benzo[b]chrysene, 1,2:6,7-Dibenzophenanthrene, 2,3-Benzochrysene, 2,3:7,8-Dibenzophenanthrene, 3,4-Benzotetracene, 3,4-Benzotetraphene, Benzo[c]tetraphene, NSC 89274, Naphth[2,1-a]anthracene
InChI formel InChI=1S/C22H14/c1-2-7-17-14-22-18(13-16(17)6-1)10-12-20-19-8-4-3-5-15(19)9-11-21(20)22/h1-14H
LEDER c1ccc2cc3c(ccc4c5ccccc5ccc34)cc2c1
IUPAC-namn Benzo[b]chrysene
Molekylvikt (g/mol) 278.3466
Formel vikt 278.1096