missing translation for 'onlineSavingsMsg'
Läs mer

Benzo[b]thiophene-2-methanol, 97%, Thermo Scientific™

Produktkod. p-7029987
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11431770

Brand: Thermo Scientific Alfa Aesar H27512.03

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Benzo[b]thiophene-2-methano is an important intermediates and reagents in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17890-56-1
Molekylformel C9H8OS
Molekylvikt (g/mol) 164.222
MDL-nummer MFCD02682004
InChI-nyckel SUHKSQTXXZQEBH-UHFFFAOYSA-N
Synonym 2-hydroxymethyl benzo b thiophene, benzo b thiophene-2-methanol, benzo b thiophen-2-ylmethanol, benzo b thiophen-2-yl-methanol, 1-benzothiophen-2-yl methanol, 1-benzothien-2-ylmethanol, 2-hydroxymethyl benzothiophene, benzothiophen-2-methanol, hydroxymethylbenzothiophene, benzothiophene-2-methanol
PubChem CID 2776335
IUPAC-namn 1-benzothiophen-2-ylmethanol
LEDER C1=CC=C2C(=C1)C=C(S2)CO

Specifications

Kemiskt namn eller material Benzo[b]thiophene-2-methanol
Smältpunkt 98°C to 101°C
Kvantitet 1 g
Formel vikt 164.22
Procent renhet 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.