missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7034209
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
171364-86-6
C14H17BO2S
260.158
MFCD11110552
LTSGSDOTQABJMA-UHFFFAOYSA-N
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
10658979
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
This item is not returnable. View return policy
171364-86-6
C14H17BO2S
260.158
MFCD11110552
LTSGSDOTQABJMA-UHFFFAOYSA-N
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
10658979
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 260.16
Percent Purity 95%
Chemical Name or Material Benzo[b]thiophene-3-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'