missing translation for 'onlineSavingsMsg'
Läs mer

Benzo[c]phenanthrene, TRC

Produktkod. 30135942
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135942 5 mg
30132517 50 mg
17990945 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30135942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1219005MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 195-19-7
Molekylformel C18 H12
Molekylvikt (g/mol) 228.29
Synonym Benzo[c]phenanthrene (8CI, 9CI, ACI), 3,4-Benzophenanthrene, Benzo[e]phenanthrene, Tetrahelicene
IUPAC-namn benzo[c]phenanthrene
LEDER c1ccc2c(c1)ccc3ccc4ccccc4c23

Specifica

Kemiskt namn eller material Benzo(c)phenanthrene
Analyt- eller komponentnamn Benzo(c)phenanthrene
Frakt skick Room Temperature
InChI formel InChI=1S/C18H12/c1-3-7-16-13(5-1)9-11-15-12-10-14-6-2-4-8-17(14)18(15)16/h1-12H
Lösningstyp Neat
Industry Type Forensics
Kvantitet 5 mg
Formel vikt 228.0939
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.