missing translation for 'onlineSavingsMsg'
Läs mer

Benzo[d]isoxazol-3-yl-methanesulfonyl Chloride, Technical Grade, TRC

Produktkod. 30345282
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30345282 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30345282 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB204100500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Benzo[d]isoxazol-3-yl-methanesulfonyl chloride
CAS 73101-65-2
Molekylformel C8 H6 Cl N O3 S
Kvantitet 500 mg
Synonym 1,2-Benzisoxazole-3-methanesulfonyl chloride, Benzo[d]isoxazol-3-ylmethanesulfonyl chloride
InChI formel InChI=1S/C8H6ClNO3S/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2
LEDER ClS(=O)(=O)Cc1noc2ccccc12
IUPAC-namn 1,2-benzoxazol-3-ylmethanesulfonyl chloride
Molekylvikt (g/mol) 231.66
Formel vikt 230.9757
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.