Läs mer
Benzo[k]fluoranthene, TRC
Produktkod. 18081830
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18081830 1 g
18013120 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18081830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2035601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 207-08-9
Molekylformel C20 H12
Molekylvikt (g/mol) 252.31
Synonym Benzo[k]fluoranthene, 11,12-Benzofluoranthene, 2,3,1',8'-Binaphthylene, 8,9-Benzfluoranthene, 8,9-Benzofluoranthene, Dibenzo[b,jk]fluorene
IUPAC-namn benzo[k]fluoranthene
LEDER c1ccc2cc3c(cc2c1)c4cccc5cccc3c45
Specifikationer
Kemiskt namn eller material Benzo(k)fluoranthene
Analyt- eller komponentnamn Benzo(k)fluoranthene
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12H
Lösningstyp Neat
Industry Type Forensics
Kvantitet 1 g
Formel vikt 252.0939
Procent renhet >95%