missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzofuroxan, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 67°C to 71°C |
Quantity | 10 g |
Beilstein | 125112 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 136.11 |
Percent Purity | 98% |
Chemical Name or Material | Benzofuroxan |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'