Läs mer
Benzoic-13C Acid, TRC
Produktkod. 30328758
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30328758 250 mg
1 options
Produktkod. 30328758 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB203955250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzoic-13C Acid
CAS 3880-99-7
Molekylformel C613CH6O2
Kvantitet 250 mg
Synonym Benzoic-carboxy-13C acid (6CI, 7CI, 8CI, 9CI, ACI), Benzoic-13C acid, Benzoic-1'-13C acid, α-13C-Benzoic acid, Benzoic Acid-α-13C
InChI formel InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)/i7+1
LEDER C1=CC=C(C=C1)[13C](=O)O
IUPAC-namn benzoic acid
Molekylvikt (g/mol) 123.11