Läs mer
Bensoesyra Acyl-Beta-D-glukuronid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzoic Acid Acyl-beta-D-glucuronide |
| CAS | 19237-53-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C13 H14 O8 |
| Kvantitet | 10 mg |
| Synonym | Benzoic Acid Acyl-β-D-glucuronide, Glucopyranuronic Acid 1-Benzoate, β-D-Glucopyranuronic Acid 1-Benzoate, Benzoic Acid glucuronide, Benzoic Acid β-D-glucuronide, Benzoyl Glucuronide, Benzoyl-β-D-glucuronide |
| InChI formel | InChI=1S/C13H14O8/c14-7-8(15)10(11(17)18)20-13(9(7)16)21-12(19)6-4-2-1-3-5-6/h1-5,7-10,13-16H,(H,17,18)/t7-,8-,9+,10-,13-/m0/s1 |
| LEDER | O[C@@H]1[C@@H](O)[C@H](OC(=O)c2ccccc2)O[C@@H]([C@H]1O)C(=O)O |
| IUPAC-namn | (2S,3S,4S,5R,6S)-6-bensoyloxi-3,4,5-trihydroxyoxan-2-karboxylsyra |
| Molekylvikt (g/mol) | 298.2455 |
| Visa mer |