Läs mer
Benzoic Acid, Supelco™
Beskrivning
Benzoic acid is a volumetric standard produced under rigorous conditions and qualified with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzoic Acid |
| CAS | 65-85-0 |
| Smältpunkt | 121°C to 123°C |
| Kokpunkt | 249°C/1013 hPa |
| Densitet | 1.321 g/cm3 at 20°C |
| Kvantitet | 60 g |
| MDL-nummer | MFCD00002398 |
| Synonym | Benzoic acid, Phenylformic acid, Benzene carboxylic acid, Benzoic acid |
| Molekylvikt (g/mol) | 122.12 |