Learn More
Benzoin methyl ether, 97%, Thermo Scientific Chemicals
Catalog Number p-7080669
Click to view available options
Quantity:
25 g
100 g
500 g
3524-62-7
C15H14O2
226.275
MFCD00008492
BQZJOQXSCSZQPS-UHFFFAOYSA-N
benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
98097
2-methoxy-1,2-diphenylethanone
COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
3524-62-7
C15H14O2
226.275
MFCD00008492
BQZJOQXSCSZQPS-UHFFFAOYSA-N
benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
98097
2-methoxy-1,2-diphenylethanone
COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.
Solubility
Soluble in methanol.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 46°C to 50°C
Density 1.128
Boiling Point 188°C to 189°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2052112
Merck Index 14,1093
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 97%
Chemical Name or Material Benzoin methyl ether