missing translation for 'onlineSavingsMsg'
Läs mer

Benzoisothiazol-3-one-13C6, TRC

Produktkod. 17970965
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.5 mg
1 mg
10 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17970965 0.5 mg 0.5mg
17933465 1 mg -
17923915 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17970965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2040370.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329616-16-1
Molekylformel 13C6 C H5 N O S
Molekylvikt (g/mol) 157.14
Synonym 1,2-Benzisothiazol-3(2H)-one-13C6, 3-Hydroxy-1,2-benzisothiazole-13C6, Acticide BIT-13C6, Apizas AP-DS-13C6, BIT-13C6, Benzisothiazolone-13C6, Benzo[d]isothiazol
IUPAC-namn 1,2-benzothiazol-3-one
LEDER O=C1NS[13c]2[13cH][13cH][13cH][13cH][13c]12

Specifications

Kemiskt namn eller material 1,2-Benzisothiazol-3(2H)-one (ring-13C6)
Anteckningar om renhetsgrad HPLC
Kvantitet 0.5 mg
InChI formel InChI=1S/C7H5NOS/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 157.0293
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.