missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, for HPLC, Thermo Scientific Chemicals

Catalog Number p-4813337
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
100-47-0
C7H5N
103.12
JFDZBHWFFUWGJE-UHFFFAOYSA-N
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
7505
CHEBI:27991
benzonitrile
C1=CC=C(C=C1)C#N
100-47-0
C7H5N
103.12
JFDZBHWFFUWGJE-UHFFFAOYSA-N
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
7505
CHEBI:27991
benzonitrile
C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -13.0°C
Density 1.0100g/mL
Boiling Point 188.0°C to 191.0°C
Flash Point 71°C
Packaging Glass bottle
Linear Formula C6H5CN
Refractive Index 1.5270 to 1.5290
Quantity 100 mL
Merck Index 15,1099
Specific Gravity 1.01
Solubility Information Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Viscosity 1.24 mPa.s (25°C)
Formula Weight 103.12
Grade HPLC
Chemical Name or Material Benzonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'