Läs mer
Benzophenone-13C, TRC
Produktkod. 30012666
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012666 50 mg
30005476 5 mg
30010078 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB42800350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32488-48-5
Molekylformel 13C C12 H10 O
Molekylvikt (g/mol) 183.21
Synonym Methanone-13C, diphenyl- (9CI, ACI), Diphenylmethanone-13C (ACI), Benzophenone-carbonyl-13C, Carbonyl-13C-benzophenone, Benzophenone-13C1 (carbonyl-13C), Benzophenone-13C (carbonyl-13C)
IUPAC-namn diphenyl(1^{13}C)methanone
LEDER O=[13C](c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Benzophenone-13C
Kvantitet 50 mg
InChI formel InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i13+1
Formel vikt 183.0765