Läs mer
Benzotriazole-5-carboxylic Acid, TRC
Produktkod. 30328970
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30328970 500 mg
1 options
Produktkod. 30328970 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB207555500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1H-Benzotriazole-5-carboxylic Acid
CAS 23814-12-2
Molekylformel C7 H5 N3 O2
Kvantitet 500 mg
Synonym 1H-Benzotriazole-5-carboxylic acid (8CI,9CI), 1,2,3-Benzotriazole-5-carboxylic acid, 1H-1,2,3-Benzotriazole-5-carboxylic acid, 1H-1,2,3-Benzotriazole-6-carboxylic acid, 1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 1H-Benzo[d][1,2,3]triazole-6-carboxylic acid, 3H-Benzotriazole-5-carboxylic acid, 5-Carboxybenzotriazole, 5-Carboxylic acid benzotriazole, Benzotriazole-5-carboxylic acid, 1H-Benzotriazole-6-carboxylic acid, 1H-Benzotriazole-5-carboxylic Acid
InChI formel InChI=1S/C7H5N3O2/c11-7(12)4-1-2-5-6(3-4)9-10-8-5/h1-3H,(H,11,12)(H,8,9,10)
LEDER OC(=O)c1ccc2nn[nH]c2c1
IUPAC-namn 3H-benzotriazole-5-carboxylic acid
Molekylvikt (g/mol) 163.134
Formel vikt 163.038