Läs mer
Benzoylaconine, TRC
Produktkod. 30136476
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136476 2.5 mg
30133033 10 mg
30005617 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2077602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 466-24-0
Molekylformel C32H45NO10
Molekylvikt (g/mol) 603.7
Synonym 14-Benzoylaconine, 14-O-Benzoylaconine, Aconine 14-Benzoate, Benzaconine, Benzoylaconine, Isaconitine, Pikraconitin;
IUPAC-namn [(1S,2R,3R,4R,5R,6S,7S,8R,9R,10R,13R,14R,16S,17S,18R)-11-ethyl-5,7,8,14-tetrahydroxy-6,16,18-trimethoxy-13-(methoxymethyl)-11-azahexacyclo[7.7.2.12,5.01,10.03,8.013,17]nonadecan-4-yl] benzoate
LEDER CCN1C[C@]2(COC)[C@@H](C[C@@H](C34[C@@H]5C[C@]6([C@@H]([C@@H]5[C@@](C([C@@H]([C@H]23)OC)C41)([C@H]([C@@H]6OC)O)O)OC(=O)C7=CC=CC=C7)O)OC)O
Specifikationer
Kemiskt namn eller material Benzoylaconine
Kvantitet 2.5 mg
InChI formel InChI=1S/C32H45NO10/c1-6-33-14-29(15-39-2)18(34)12-19(40-3)31-17-13-30(37)26(43-28(36)16-10-8-7-9-11-16)20(17)32(38,25(35)27(30)42-5)21(24(31)33)22(41-4)23(29)31/h7-11,17-27,34-35,37-38H,6,12-15H2,1-5H3/t17-,18-,19+,20-,21+,22+,23-,24-,25+,26-,27+,29+,30-,31+,32-/m1/s1