Läs mer
Benzyl 2,3,4-Tri-O-benzyl-D-glucuronate Acetate, TRC
Produktkod. 30338532
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30338532 25 mg
1 options
Produktkod. 30338532 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB31589025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 2,3,4-Tri-O-benzyl-D-glucuronate Acetate
CAS 4550-93-0
Molekylformel C36H36O8
Kvantitet 25 mg
Synonym 2,3,4-Tris-O-(phenylmethyl)-D-glucopyranuronic Acid Phenylmethyl Ester Acetate, 2,3,4-Tris-O-(phenylmethyl)-D-glucopyranuronic Acid Benzyl Ester Acetate
InChI formel InChI=1S/C36H36O8/c1-26(37)43-36-34(41-24-29-18-10-4-11-19-29)32(40-23-28-16-8-3-9-17-28)31(39-22-27-14-6-2-7-15-27)33(44-36)35(38)42-25-30-20-12-5-13-21-30/h2-21,31-34,36H,22-25H2,1H3/t31-,32-,33-,34+,36?/m0/s1
LEDER CC(=O)OC1O[C@@H]([C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@H]1OCc4ccccc4)C(=O)OCc5ccccc5
IUPAC-namn benzyl (2S,3S,4S,5R)-6-acetyloxy-3,4,5-tris(phenylmethoxy)oxane-2-carboxylate
Molekylvikt (g/mol) 596.67
Formel vikt 596.241