Läs mer
Benzyl 2-Acetamido-2-deoxy-3-O-(Beta-D-galactopyranosyl)-Alpha-D-glucopyranoside, TRC
Produktkod. 30338843
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30338843 10 mg
1 options
Produktkod. 30338843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB21253210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 2-Acetamido-2-deoxy-3-O-beta-D-galactopyranosyl-alpha-D-glucopyranoside
CAS 50692-66-5
Molekylformel C21 H31 N O11
Kvantitet 10 mg
Synonym α-D-Glucopyranoside, phenylmethyl 2-(acetylamino)-2-deoxy-3-O-β-D-galactopyranosyl-, Glucopyranoside, benzyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl- (7CI), Phenylmethyl 2-(acetylamino)-2-deoxy-3-O-β-D-galactopyranosyl-α-D-glucopyranoside, Benzyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl-α-D-glucopyranoside
InChI formel InChI=1S/C21H31NO11/c1-10(25)22-14-19(33-21-18(29)17(28)15(26)12(7-23)32-21)16(27)13(8-24)31-20(14)30-9-11-5-3-2-4-6-11/h2-6,12-21,23-24,26-29H,7-9H2,1H3,(H,22,25)/t12-,13-,14-,15+,16-,17+,18-,19-,20+,21+/m1/s1
LEDER CC(=O)N[C@H]1[C@@H](OCc2ccccc2)O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O
IUPAC-namn N-[(2S,3R,4R,5S,6R)-5-hydroxy-6-(hydroxymethyl)-2-phenylmethoxy-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
Molekylvikt (g/mol) 473.47
Formel vikt 473.1897