Läs mer
Benzyl 2-Acetamido-2-deoxy-4,6-O-benzylidene-alpha-D-galactopyranoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzyl 2-Acetamido-2-deoxy-4,6-O-benzylidene-alpha-D-galactopyranoside |
| CAS | 3554-91-4 |
| Molekylformel | C22 H25 N O6 |
| Kvantitet | 100 mg |
| Synonym | α-D-Galactopyranoside, phenylmethyl 2-(acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-, Galactopyranoside, benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy- (7CI), Galactopyranoside, benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-, α-D- (8CI), Phenylmethyl 2-(acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-α-D-galactopyranoside, Pyrano[3,2-d]-1,3-dioxin, α-D-galactopyranoside deriv., Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-α-D-galactopyranoside, Benzyl 2-acetamido-2-deoxy-4,6-O-benzylidene-α-D-galactopyranoside |
| InChI formel | InChI=1S/C22H25NO6/c1-14(24)23-18-19(25)20-17(13-27-21(29-20)16-10-6-3-7-11-16)28-22(18)26-12-15-8-4-2-5-9-15/h2-11,17-22,25H,12-13H2,1H3,(H,23,24)/t17-,18-,19-,20+,21?,22+/m1/s1 |
| LEDER | CC(=O)N[C@H]1[C@@H](OCc2ccccc2)O[C@@H]3COC(O[C@@H]3[C@@H]1O)c4ccccc4 |
| IUPAC-namn | N-[(4aR,6S,7R,8R,8aR)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide |
| Molekylvikt (g/mol) | 399.44 |
| Formel vikt | 399.1682 |