Läs mer
Benzyl 2-Acetamido-3-O-allyl-4,6-O-benzylidene-2-deoxy-Alpha-D-glucopyranoside, TRC
Produktkod. 30342930
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30342930 1 g
30338370 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30342930 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2101001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 2-Acetamido-3-O-allyl-4,6-O-benzylidene-2-deoxy-a-D-g
CAS 60920-72-1
Molekylformel C25 H29 N O6
Kvantitet 1 g
InChI formel InChI=1S/C25H29NO6/c1-3-14-28-23-21(26-17(2)27)25(29-15-18-10-6-4-7-11-18)31-20-16-30-24(32-22(20)23)19-12-8-5-9-13-19/h3-13,20-25H,1,14-16H2,2H3,(H,26,27)/t20-,21-,22-,23-,24?,25+/m1/s1
LEDER CC(=O)N[C@H]1[C@@H](OCc2ccccc2)O[C@@H]3COC(O[C@H]3[C@@H]1OCC=C)c4ccccc4
IUPAC-namn N-[(4aR,6S,7R,8R,8aS)-2-phenyl-6-phenylmethoxy-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Molekylvikt (g/mol) 439.5
Formel vikt 439.1995