Läs mer
Benzyl 2-Acetamido-4,6-O-Benzylidene-2-Deoxy-Beta-D-Glucopyranoside, TRC
Produktkod. 30344613
Change view
Kvantitet:
2 g
Produktkod. Kvantitet
30344613 2 g
1 options
Produktkod. 30344613 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2110012G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 2-Acetamido-4,6-O-Benzylidene-2-Deoxy-b-D-Glucopyrano
CAS 13343-61-8
Molekylformel C22 H25 N O6
Kvantitet 2 g
Synonym β-D-Glucopyranoside, phenylmethyl 2-(acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-, Glucopyranoside, benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-, β-D- (8CI), Phenylmethyl 2-(acetylamino)-2-deoxy-4,6-O-(phenylmethylene)-β-D-glucopyranoside, Pyrano[3,2-d]-1,3-dioxin, β-D-glucopyranoside deriv., Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside
InChI formel InChI=1S/C22H25NO6/c1-14(24)23-18-19(25)20-17(13-27-21(29-20)16-10-6-3-7-11-16)28-22(18)26-12-15-8-4-2-5-9-15/h2-11,17-22,25H,12-13H2,1H3,(H,23,24)/t17-,18-,19-,20-,21?,22-/m1/s1
LEDER CC(=O)N[C@H]1[C@H](OCc2ccccc2)O[C@@H]3COC(O[C@H]3[C@@H]1O)c4ccccc4
IUPAC-namn N-[(4aR,6R,7R,8R,8aS)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Molekylvikt (g/mol) 399.44
Formel vikt 399.1682