missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoacetate, 96%, Thermo Scientific Chemicals

Catalog Number p-65643
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
5437-45-6
C9H9BrO2
229.08
MFCD00000190
JHVLLYQQQYIWKX-UHFFFAOYSA-N
benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
62576
benzyl 2-bromoacetate
C1=CC=C(C=C1)COC(=O)CBr
This item is not returnable. View return policy
5437-45-6
C9H9BrO2
229.08
MFCD00000190
JHVLLYQQQYIWKX-UHFFFAOYSA-N
benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
62576
benzyl 2-bromoacetate
C1=CC=C(C=C1)COC(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 166.0°C to 170.0°C (22.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.5450
Quantity 50 g
Specific Gravity 1.44
Solubility Information Solubility in water: insoluble.
Formula Weight 229.08
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Benzyl 2-bromoacetate, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'