Läs mer
Benzyl 3-Hydroxypropionate, TRC
Produktkod. 30340559
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30340559 1 g
1 options
Produktkod. 30340559 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2799651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 3-Hydroxypropionate
CAS 14464-10-9
Molekylformel C10H12O3
Kvantitet 1 g
Synonym 3-Hydroxy-propanoic Acid Phenylmethyl Ester, Hydracrylic Acid Benzyl Ester, Benzyl 3-Hydroxypropanoate
InChI formel InChI=1S/C10H12O3/c11-7-6-10(12)13-8-9-4-2-1-3-5-9/h1-5,11H,6-8H2
LEDER OCCC(=O)OCc1ccccc1
IUPAC-namn benzyl 3-hydroxypropanoate
Molekylvikt (g/mol) 180.2
Formel vikt 180.0786