Läs mer
Benzyl 4-Nitrophenyl Carbonate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzyl 4-Nitrophenyl Carbonate |
| CAS | 13795-24-9 |
| Molekylformel | C14H11NO5 |
| Kvantitet | 5 g |
| Synonym | Carbonic Acid 4-Nitrophenyl Phenylmethyl Ester, Carbonic Acid Benzyl p-Nitrophenyl Ester, 4-Nitrophenyl Benzyl Carbonate, Benzyl p-Nitrophenyl Carbonate, NSC 171047, p-Nitrophenyl Benzyl Carbonate |
| InChI formel | InChI=1S/C14H11NO5/c16-14(19-10-11-4-2-1-3-5-11)20-13-8-6-12(7-9-13)15(17)18/h1-9H,10H2 |
| LEDER | [O-][N+](=O)c1ccc(OC(=O)OCc2ccccc2)cc1 |
| IUPAC-namn | benzyl (4-nitrophenyl) carbonate |
| Molekylvikt (g/mol) | 273.24 |
| Formel vikt | 273.0637 |