missing translation for 'onlineSavingsMsg'
Läs mer

Benzyl 4-Nitrophenyl Carbonate, TRC

Produktkod. 30340420
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30340420 50 g
30330941 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30340420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB28548550G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Benzyl 4-Nitrophenyl Carbonate
CAS 13795-24-9
Molekylformel C14H11NO5
Kvantitet 50 g
Synonym Carbonic Acid 4-Nitrophenyl Phenylmethyl Ester, Carbonic Acid Benzyl p-Nitrophenyl Ester, 4-Nitrophenyl Benzyl Carbonate, Benzyl p-Nitrophenyl Carbonate, NSC 171047, p-Nitrophenyl Benzyl Carbonate
InChI formel InChI=1S/C14H11NO5/c16-14(19-10-11-4-2-1-3-5-11)20-13-8-6-12(7-9-13)15(17)18/h1-9H,10H2
LEDER [O-][N+](=O)c1ccc(OC(=O)OCc2ccccc2)cc1
IUPAC-namn benzyl (4-nitrophenyl) carbonate
Molekylvikt (g/mol) 273.24
Formel vikt 273.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.