Läs mer
Benzyl 6-Hydroxyhexanoate, TRC
Produktkod. 30338807
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30338807 500 mg
1 options
Produktkod. 30338807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB279250500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 6-Hydroxyhexanoate
CAS 125878-06-0
Molekylformel C13 H18 O3
Kvantitet 500 mg
Synonym Benzyl 6-Hydroxycaproate, 6-Hydroxy-hexanoic Acid Phenylmethyl Ester, 2,5-Dioxo-1-pyrrolidinyl 4-Azidobutanoate, (2,5-Dioxopyrrolidin-1-yl) 4-Azidobutanoate, 2,5-Dioxopyrrolidin-1-yl 4-azidobutanoate
InChI formel InChI=1S/C13H18O3/c14-10-6-2-5-9-13(15)16-11-12-7-3-1-4-8-12/h1,3-4,7-8,14H,2,5-6,9-11H2
LEDER O=C(OCC=1C=CC=CC1)CCCCCO
IUPAC-namn benzyl 6-hydroxyhexanoate
Molekylvikt (g/mol) 222.28022
Formel vikt 222.1256