Läs mer
Benzyl 6-Methyloctanoate, TRC
Produktkod. 30334779
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30334779 500 mg
1 options
Produktkod. 30334779 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB285723500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl 6-Methyloctanoate
CAS 2383054-95-1
Molekylformel C16H24O2
Kvantitet 500 mg
Synonym Octanoic acid, 6-methyl-, phenylmethyl ester
InChI formel InChI=1S/C16H24O2/c1-3-14(2)9-7-8-12-16(17)18-13-15-10-5-4-6-11-15/h4-6,10-11,14H,3,7-9,12-13H2,1-2H3
LEDER O=C(CCCCC(C)CC)OCC1=CC=CC=C1
IUPAC-namn Benzyl 6-Methyloctanoate
Molekylvikt (g/mol) 248.36
Formel vikt 248.18