Läs mer
Benzyl beta-D-Glucopyranoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzyl beta-D-Glucopyranoside |
| CAS | 4304-12-5 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C13 H18 O6 |
| Kvantitet | 5 g |
| Synonym | β-D-Glucopyranoside, phenylmethyl, Glucopyranoside, benzyl, β-D- (7CI,8CI), Phenylmethyl β-D-glucopyranoside, Benzyl alcohol glucoside, Benzyl glucopyranoside, Benzyl β-D-glucopyranoside, Benzyl β-D-glucoside, Benzyl-O-β-D-glucopyranoside |
| InChI formel | InChI=1S/C13H18O6/c14-6-9-10(15)11(16)12(17)13(19-9)18-7-8-4-2-1-3-5-8/h1-5,9-17H,6-7H2/t9-,10-,11+,12-,13-/m1/s1 |
| LEDER | OC[C@H]1O[C@@H](OCc2ccccc2)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC-namn | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-phenylmethoxyoxane-3,4,5-triol |
| Molekylvikt (g/mol) | 270.28 |
| Visa mer |