Läs mer
Benzyl beta-D-Glucopyranosiduronic Acid, TRC
Produktkod. 30329832
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30329832 100 mg
1 options
Produktkod. 30329832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB276780100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl beta-D-Glucopyranosiduronic Acid
CAS 5285-02-9
Molekylformel C13H16O7
Kvantitet 100 mg
Synonym Benzyl Alcohol Glucuronide, Benzyl β-D-Glucopyranosiduronic Acid
InChI formel InChI=1S/C13H16O7/c14-8-9(15)11(12(17)18)20-13(10(8)16)19-6-7-4-2-1-3-5-7/h1-5,8-11,13-16H,6H2,(H,17,18)/t8-,9-,10+,11-,13+/m0/s1
LEDER C1=CC=C(C=C1)CO[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O
IUPAC-namn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-phenylmethoxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 284.26
Formel vikt 284.0896