Läs mer
Benzyl Beta-D-Lactoside, TRC
Produktkod. 30330953
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30330953 100 mg
30332875 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30330953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB281625100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl beta-D-lactoside
CAS 18404-72-3
Molekylformel C19 H28 O11
Kvantitet 100 mg
InChI formel InChI=1S/C19H28O11/c20-6-10-12(22)13(23)15(25)19(28-10)30-17-11(7-21)29-18(16(26)14(17)24)27-8-9-4-2-1-3-5-9/h1-5,10-26H,6-8H2/t10-,11-,12+,13+,14-,15-,16-,17-,18-,19+/m1/s1
LEDER OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OCc3ccccc3)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
IUPAC-namn (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-phenylmethoxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvikt (g/mol) 432.42
Formel vikt 432.1632