Läs mer
Benzyl Butyl Phthalate, TRC
Produktkod. 30178959
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30178959 1 g
1 options
Produktkod. 30178959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2335551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phthalic acid, benzylbutyl ester
CAS 85-68-7
Analyt- eller komponentnamn Phthalic acid, benzylbutyl ester
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
Molekylformel C19 H20 O4
Synonym 1,2-Benzenedicarboxylic acid, 1-butyl 2-(phenylmethyl) ester, 1,2-Benzenedicarboxylic acid, butyl phenylmethyl ester (9CI), Phthalic acid, benzyl butyl ester (6CI,8CI), 2-O-Benzyl 1-O-butyl benzene-1,2-dicarboxylate, BBP, Benzyl butyl phthalate, Benzyl n-butyl phthalate, Butyl benzyl phthalate, Diacizer D 160, Ketjenflex 160, NSC 71001, Palatinol BB, S 160, Sant 160, Santicizer 160, Santicizer C 160, Santicizer S 106, Santicizer S 160, Sicol 160, Spatozoate, Unimoll BB, n-Butyl benzyl phthalate, n-Butyl benzyl phthalate diester, Phthalic acid benzyl butyl ester
InChI formel InChI=1S/C19H20O4/c1-2-3-13-22-18(20)16-11-7-8-12-17(16)19(21)23-14-15-9-5-4-6-10-15/h4-12H,2-3,13-14H2,1H3
Lösningstyp Neat
LEDER CCCCOC(=O)c1ccccc1C(=O)OCc2ccccc2
IUPAC-namn 2-O-benzyl 1-O-butyl benzene-1,2-dicarboxylate
Industry Type Forensics
Molekylvikt (g/mol) 312.36
Kvantitet 1 g
Formel vikt 312.1362
Procent renhet >95%