Läs mer
Benzyl Butyl Phthalate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, benzylbutyl ester |
| CAS | 85-68-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C19 H20 O4 |
| Kvantitet | 10 g |
| Synonym | 1,2-Benzenedicarboxylic acid, 1-butyl 2-(phenylmethyl) ester, 1,2-Benzenedicarboxylic acid, butyl phenylmethyl ester (9CI), Phthalic acid, benzyl butyl ester (6CI,8CI), 2-O-Benzyl 1-O-butyl benzene-1,2-dicarboxylate, BBP, Benzyl butyl phthalate, Benzyl n-butyl phthalate, Butyl benzyl phthalate, Diacizer D 160, Ketjenflex 160, NSC 71001, Palatinol BB, S 160, Sant 160, Santicizer 160, Santicizer C 160, Santicizer S 106, Santicizer S 160, Sicol 160, Spatozoate, Unimoll BB, n-Butyl benzyl phthalate, n-Butyl benzyl phthalate diester, Phthalic acid benzyl butyl ester |
| InChI formel | InChI=1S/C19H20O4/c1-2-3-13-22-18(20)16-11-7-8-12-17(16)19(21)23-14-15-9-5-4-6-10-15/h4-12H,2-3,13-14H2,1H3 |
| LEDER | CCCCOC(=O)c1ccccc1C(=O)OCc2ccccc2 |
| IUPAC-namn | 2-O-benzyl 1-O-butyl benzene-1,2-dicarboxylate |
| Molekylvikt (g/mol) | 312.36 |
| Visa mer |