Läs mer
Benzyl Cyanomethylcarbamate, TRC
Produktkod. 30341098
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30341098 500 mg
1 options
Produktkod. 30341098 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB130295500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl Cyanomethylcarbamate
CAS 3589-41-1
Molekylformel C10H10N2O2
Kvantitet 500 mg
InChI formel InChI=1S/C10H10N2O2/c11-6-7-12-10(13)14-8-9-4-2-1-3-5-9/h1-5H,7-8H2,(H,12,13)
LEDER O=C(NCC#N)OCc1ccccc1
IUPAC-namn benzyl N-(cyanomethyl)carbamate
Molekylvikt (g/mol) 190.19
Formel vikt 190.0742