Läs mer
Benzyl D-Glucopyranoside (alpha & beta Mixture) (>90%), TRC
Produktkod. 30347024
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30347024 5 g
1 options
Produktkod. 30347024 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2767505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl D-Glucopyranoside (An alpha-beta mixture)
CAS 34246-23-6
Molekylformel C13H18O6
Kvantitet 5 g
Synonym Phenylmethyl D-Glucopyranoside (alpha & beta Mixture)
InChI formel InChI=1S/C13H18O6/c14-6-9-10(15)11(16)12(17)13(19-9)18-7-8-4-2-1-3-5-8/h1-5,9-17H,6-7H2/t9-,10-,11+,12-,13?/m1/s1
LEDER O(CC1=CC=CC=C1)C2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O
IUPAC-namn (2R,3S,4S,5R)-2-(hydroxymethyl)-6-phenylmethoxyoxane-3,4,5-triol
Molekylvikt (g/mol) 270.28
Formel vikt 270.1103