Läs mer
Benzyl D-Glucuronate, TRC
Produktkod. 30341982
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30341982 100 mg
1 options
Produktkod. 30341982 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB279800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl D-Glucuronate
CAS 135970-30-8
Molekylformel C13 H16 O7
Kvantitet 100 mg
Synonym D-Glucuronic acid, phenylmethyl ester, Benzyl D-glucuronate
InChI formel InChI=1S/C13H16O7/c14-8-9(15)11(20-12(17)10(8)16)13(18)19-6-7-4-2-1-3-5-7/h1-5,8-12,14-17H,6H2/t8-,9-,10+,11-,12?/m0/s1
LEDER OC1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)OCc2ccccc2
IUPAC-namn benzyl (2S,3S,4S,5R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
Molekylvikt (g/mol) 284.26
Formel vikt 284.0896