missing translation for 'onlineSavingsMsg'
Learn More

Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-3212406
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
57443-18-2
C11H15O5P
258.21
MFCD01872507
QYLGNJMIOVHLQQ-UHFFFAOYSA-N
benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
2773759
benzyl 2-dimethoxyphosphorylacetate
COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
This item is not returnable. View return policy
57443-18-2
C11H15O5P
258.21
MFCD01872507
QYLGNJMIOVHLQQ-UHFFFAOYSA-N
benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
2773759
benzyl 2-dimethoxyphosphorylacetate
COP(=O)(CC(=O)OCC1=CC=CC=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Brown
Boiling Point 148.0°C to 150.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)CH2C(O)OCH2C5H5
Refractive Index 1.5050 to 1.5070
Quantity 5 g
Formula Weight 258.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl dimethyl phosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl dimethyl phosphonoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'