missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ether, 99%, Thermo Scientific Chemicals

Catalog Number p-65838
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
103-50-4
C14H14O
198.26
MFCD00004780
MHDVGSVTJDSBDK-UHFFFAOYSA-N
dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
7657
CHEBI:87411
phenylmethoxymethylbenzene
C1=CC=C(C=C1)COCC2=CC=CC=C2
103-50-4
C14H14O
198.26
MFCD00004780
MHDVGSVTJDSBDK-UHFFFAOYSA-N
dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
7657
CHEBI:87411
phenylmethoxymethylbenzene
C1=CC=C(C=C1)COCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.5°C to 3.5°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 298.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5CH2)2O
Refractive Index 1.5610 to 1.5630
Quantity 1 L
Beilstein 06, 434
Merck Index 15, 1135
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and chloroform
Formula Weight 198.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl ether, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl ether
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'