missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl methyl ether, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -53°C |
Density | 0.94 |
Boiling Point | 173°C to 175°C |
Flash Point | 56°C (133°F) |
Refractive Index | 1.5015 |
Quantity | 10 g |
UN Number | UN3271 |
Merck Index | 14,1140 |
Solubility Information | Insoluble in water. |
Formula Weight | 122.17 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'