Läs mer
Benzyl N-Acetyl-4,6-O-benzylidene-Alpha-D-muramic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzyl N-Acetyl-4,6-O-benzylidene-a-D-muramic Acid |
| CAS | 2862-03-5 |
| Molekylformel | C25H29NO8 |
| Kvantitet | 100 mg |
| Synonym | α-Muramic acid, N-acetyl-1-O-(phenylmethyl)-4,6-O-(phenylmethylene)-, Glucopyranoside, benzyl 2-acetamido-4,6-O-benzylidene-3-O-(D-1-carboxyethyl)-2-deoxy- (7CI), Glucopyranoside, benzyl 2-acetamido-4,6-O-benzylidene-3-O-(D-1-carboxyethyl)-2-deoxy-, α-D- (8CI), α-D-Glucopyranoside, phenylmethyl 2-(acetylamino)-3-O-(1-carboxyethyl)-2-deoxy-4,6-O-(phenylmethylene)-, [3(R)]-, N-Acetyl-1-O-(phenylmethyl)-4,6-O-(phenylmethylene)-α-muramic acid, α-O-Benzyl-4,6-O-benzylidene-N-acetylmuramic acid, N-Acetyl-1-O-benzyl-4,6-O-(benzylidene)-α-muramic Acid |
| InChI formel | InChI=1S/C25H29NO8/c1-15(23(28)29)32-22-20(26-16(2)27)25(30-13-17-9-5-3-6-10-17)33-19-14-31-24(34-21(19)22)18-11-7-4-8-12-18/h3-12,15,19-22,24-25H,13-14H2,1-2H3,(H,26,27)(H,28,29)/t15-,19-,20-,21-,22-,24?,25+/m1/s1 |
| LEDER | C[C@@H](O[C@@H]1[C@@H](NC(=O)C)[C@@H](OCc2ccccc2)O[C@@H]3COC(O[C@@H]13)c4ccccc4)C(=O)O |
| IUPAC-namn | (2R)-2-[[(4aR,6S,7R,8R,8aS)-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy]propanoic acid |
| Molekylvikt (g/mol) | 471.5 |