Learn More
Benzyl propargyl ether, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.
Solubility
Soluble in chloroform.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.
Solubility
Soluble in chloroform.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.011 |
Boiling Point | 101°C to 101°C (18 mmHg) |
Linear Formula | C6H4CH2OCH2C≡CH |
Refractive Index | 1.52 |
Quantity | 5 g |
Solubility Information | Soluble in chloroform. |
Formula Weight | 146.19 |
Percent Purity | 97% |
Chemical Name or Material | Benzyl propargyl ether |