Läs mer
Benzyl Trisulfide, TRC
Produktkod. 30348498
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30348498 2.5 g
1 options
Produktkod. 30348498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3161502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl Trisulfide
CAS 6493-73-8
Molekylformel C14 H14 S3
Kvantitet 2.5 g
Synonym Bis(phenylmethyl)trisulfide, Dibenzyl Trisulfide, DTS
InChI formel InChI=1S/C14H14S3/c1-3-7-13(8-4-1)11-15-17-16-12-14-9-5-2-6-10-14/h1-10H,11-12H2
LEDER C(SSSCc1ccccc1)c2ccccc2
IUPAC-namn (benzyltrisulfanyl)methylbenzene
Molekylvikt (g/mol) 278.46
Formel vikt 278.0258