Läs mer
(Benzyloxy)(methyl)amine, TRC
Produktkod. 30332300
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30332300 500 mg
1 options
Produktkod. 30332300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB411713500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Benzyloxy)(methyl)amine
CAS 22513-22-0
Molekylformel C8H11NO
Kvantitet 500 mg
Synonym N-(Phenylmethoxy)methanamine, (Benzyloxy)(methyl)amine, N-Benzyloxy-N-methylamine, N-Methyl-O-benzylhydroxylamine, O-Benzyl-N-methylhydroxylamine
InChI formel InChI=1S/C8H11NO/c1-9-10-7-8-5-3-2-4-6-8/h2-6,9H,7H2,1H3
LEDER CNOCc1ccccc1
IUPAC-namn N-phenylmethoxymethanamine
Molekylvikt (g/mol) 137.17
Formel vikt 137.0841