missing translation for 'onlineSavingsMsg'
Läs mer

Benzylpenicillin Benzathine, LoGiCal

Produktkod. 30460429
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460429 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30460429 Leverantör LoGiCal Leverantörsnummer LGCFOR0513.00

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Benzathine benzylpenicillin
CAS 1538-09-6
Analyt- eller komponentnamn Benzathine benzylpenicillin
Frakt skick Ice pack (-20°C)
Molekylformel C16 H20 N2 . 2 C16 H18 N2 O4 S
Certifikat och efterlevnad ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]- (2S,5R,6R)-, compd. with N1,N2-bis(phenylmethyl)-1,2-ethanediamine (2:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-, compd. with N,N'-dibenzylethylenediamine (2:1) (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1) (9CI), Penicillin G, N,N'-dibenzylethylenediamine salt (6CI), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] (9CI), Ethylenediamine, N,N'-dibenzyl-, bis[3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] (8CI), Beacillin, Benepen, Benzacillin, Benzapen, Benzathine benzylpenicillin, Benzathine penicillin, Benzathine penicillin G, Benzethacil, Benzylpenicillin N,N'-dibenzylethylenediamine salt, Benzylpenicillin benzathine, Benzylpenicillin dibenzylethylenediamine salt, Bica-penicillin, Bicillin, Bicillin 1, Bicillin G, Bicillin I, Bicillin L-A, Cepacilina, Cillenta, Debecillin, Debecylina, Diamine penicillin, Dibencil, Dibencillin, Dibenzylethylenediamine dipenicillin G, Duropenin, Ethylenediamine, N,N'-dibenzyl-, compd. with penicillin G (1:2), Extencilline, Extenicilline, Lentopenil, Longacilina, Longicil, Megacillin suspension, Moldamin, N,N'-Dibenzylethylenediamine bis[benzyl penicillin], N,N'-Dibenzylethylenediamine salt of benzylpenicillin, Neolin, Pen-Di-Ben, Penadur, Pendepon, Penditan, Penduran, Penicillin G, compd. with N,N'-dibenzylethylenediamine (2:1), Penicom 12, Penidural, Penidure, Permapen, Strepdipen-Suspension, Tardocillin, Vicin, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- [2S-(2alpha,5alpha,6beta)]-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[[2S-(2alpha,5alpha,6beta)]-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate]
InChI formel InChI=1S/2C16H18N2O4S.C16H20N2/c2*1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h2*3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);1-10,17-18H,11-14H2/t2*11-,12+,14-;/m11./s1
Lösningstyp Neat
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(=O)O.CC4(C)S[C@@H]5[C@H](NC(=O)Cc6ccccc6)C(=O)N5[C@H]4C(=O)O.C(CNCc7ccccc7)NCc8ccccc8
IUPAC-namn N,N'-dibenzylethane-1,2-diamine;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 909.12
Kvantitet 10 mg
Formel vikt 908.3601 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.