Läs mer
Benzylpenicillin Diethylaminoethyl Ester Hydroiodide, TRC
Produktkod. 30132412
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132412 1 g
30009632 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132412 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2885901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 808-71-9
Molekylformel C22H32IN3O4S
Molekylvikt (g/mol) 561.48
Synonym 3,3-Dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 2-(Diethylamino)ethyl Ester Monohydriodide, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 2-(Diethylamino)ethyl Ester Monohydriodide, Alivin, Benzylpenicillin β-diethylaminoethyl ester hydriodide, Bronchocillin, Bronchopen, Broncopen, Deripen, Diethylaminoethyl Ester Penicillin Hydriodide, Eficillin, Ephicillin Hydriodide, Estopen, Leocillin, Mamyzin, Neo-Penil, Penethacillin, Penethamate Hydriodide, Penethecillin, Penicillin G Diethylaminoethyl Ester Hydriodide, Pulmaxil N, Pulmo 500, β-Diethylaminoethyl Benzylpenicillinate Hydriodide;
IUPAC-namn 2-(diethylamino)ethyl (2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydroiodide
LEDER I.CCN(CC)CCOC(=O)[C@@H]1N2[C@H](SC1(C)C)[C@H](NC(=O)Cc3ccccc3)C2=O
Specifikationer
Kemiskt namn eller material Benzylpenicillin Diethylaminoethyl Ester Hydroiodide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C22H31N3O4S.HI/c1-5-24(6-2)12-13-29-21(28)18-22(3,4)30-20-17(19(27)25(18)20)23-16(26)14-15-10-8-7-9-11-15;/h7-11,17-18,20H,5-6,12-14H2,1-4H3,(H,23,26);1H/t17-,18+,20-;/m1./s1
Formel vikt 561.1158
Procent renhet >95%