missing translation for 'onlineSavingsMsg'
Läs mer

Benzyltriethylammonium borohydride, 96%, Thermo Scientific™

Produktkod. 11496420
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11496420 5 g 5g
11406430 25 g 25g
11416430 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11496420 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B24952.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Benzyltriethylammonium borohydride involves in the conversion of alkenes to the anti-Markovnikov alcohols in presence of TMS chloride or TiCl4. It combines with TMS chloride, selectively reduces carboxylic acids to alcohols; esters, lactones, nitro- and chloro-substituents are unaffected. It is used as reducing reagent.

Solubility
Souble in Chloroform.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 85874-45-9
Molekylformel C13H22BN
Molekylvikt (g/mol) 203.136
MDL-nummer MFCD00191785
InChI-nyckel HQGWCEJYXHFBSA-UHFFFAOYSA-N
Synonym benzyltriethylammonium borohydride, benzyltriethylazanium ? 1-boranuidylidene, n,n,n-triethyl-benzenemethanaminium tetrahydroborate
PubChem CID 10899792
IUPAC-namn benzyl(triethyl)azanium;boron(1-)
LEDER [B-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifikationer

Kemiskt namn eller material Benzyltriethylammonium borohydride
Smältpunkt ∼146°C (decomposition)
Linjär formel (CH3CH2)3N(CH2C6H5)BH4
Kvantitet 5 g
UN-nummer UN3131
Känslighet Moisture sensitive
Löslighetsinformation Souble in Chloroform.
Formel vikt 207.17
Procent renhet 96%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.